Palbociclib isethionate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Acetyl-8-cyclopentyl-5-methyl-2-((5-(piperazin-1-yl)pyridin-2-yl)amino)pyrido[2,3-d]pyrimidin-7(8H)-one 2-hydroxyethanesulphonate
A280869-10MG
A280869-1MG
A280869-50MG
A280869-5MG
77153-812EA
64.89
USD
77153-812
77153-814
77153-816
77153-818
Palbociclib isethionate 98%
Palbociclib isethionate
Precaución: for Research Use Only
Formula:
C₂₄H₂₉N₇O₂·C₂H₆O₄S MW: 573.66 g/mol |
MDL Number:
MFCD22666589 CAS Number: 827022-33-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...