4-Amino-5-chloro-N-(1-(3-methoxypropyl)piperidin-4-yl)-2,3-dihydrobenzofuran-7-carboxamide succinate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Prucalopride succinate
A280691-10MG
A280691-1G
A280691-250MG
A280691-50MG
A280691-5G
77153-804EA
5.15
USD
77153-804
77153-806
77153-808
77153-810
77550-722
4-Amino-5-chloro-N-(1-(3-methoxypropyl)piperidin-4-yl)-2,3-dihydrobenzofuran-7-carboxamide succinate 98%
4-Amino-5-chloro-N-(1-(3-methoxypropyl)piperidin-4-yl)-2,3-dihydrobenzofuran-7-carboxamide succinate
Precaución: for Research Use Only
Formula:
C₂₂H₃₂ClN₃O₇ MW: 485.96 g/mol |
MDL Number:
MFCD00948731 CAS Number: 179474-85-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...