Methyl 6-(TrifluoroMethoxy)indole-2-carboxylate 98%

Supplier: Ambeed
Warning

A280390-100MG A280390-1G A280390-250MG
77153-798EA 53.56 USD
77153-798 77153-800 77153-802
Methyl 6-(TrifluoroMethoxy)indole-2-carboxylate 98%
Methyl 6-(TrifluoroMethoxy)indole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₈F₃NO₃
MW: 259.18 g/mol
MDL Number: MFCD12070772
CAS Number: 1154319-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR