3-(3-Fluorophenyl)prop-2-yn-1-ol 97%

Supplier: Ambeed
Warning

Synonyms: 1-(3-Fluorophenyl)-3-hydroxyprop-1-yne

A278633-100MG A278633-1G A278633-250MG
77153-780EA 20.6 USD
77153-780 77153-782 77153-784
3-(3-Fluorophenyl)prop-2-yn-1-ol 97%
3-(3-Fluorophenyl)prop-2-yn-1-ol

Precaución: for Research Use Only

Formula: C₉H₇FO
MW: 150.15 g/mol
MDL Number: MFCD04039231
CAS Number: 197239-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR