SGI-1027 ≥98%

Supplier: Ambeed
Warning

Synonyms: N-(4-((2-amino-6-methylpyrimidin-4-yl)amino)phenyl)-4-(quinolin-4-ylamino)benzamide, N-[4-[(2-Amino-6-methyl-4-pyrimidinyl)amino]phenyl]-4-(4-quinolinylamino)benzamide

A278291-10MG A278291-1MG
77153-772EA 48.41 USD
77153-772 77153-774
SGI-1027 ≥98%
SGI-1027

Precaución: for Research Use Only

Formula: C₂₇H₂₃N₇O
MW: 461.52 g/mol
MDL Number: MFCD27937047
CAS Number: 1020149-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR