3-Methyl-5-nitro-1H-pyrrolo[2,3-b]pyridine 95%

Supplier: Ambeed
Warning

A277232-100MG A277232-1G A277232-250MG
77153-758EA 81.37 USD
77153-758 77153-760 77153-762
3-Methyl-5-nitro-1H-pyrrolo[2,3-b]pyridine 95%
3-Methyl-5-nitro-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₈H₇N₃O₂
MW: 177.16 g/mol
MDL Number: MFCD20527500
CAS Number: 1288998-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR