4-Phenylcyclohexanone oxime 95+%

Supplier: Ambeed
Danger

A277223-100MG A277223-1G A277223-250MG A277223-5G
77153-750EA 19.57 USD
77153-750 77153-752 77153-754 77153-756
4-Phenylcyclohexanone oxime 95+%
4-Phenylcyclohexanone oxime

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO
MDL Number: MFCD00456899
CAS Number: 4500-20-3
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR