L(+)-Penicillamine 95%
Supplier: Ambeed
|
Danger
|
A276897-100MG
A276897-1G
A276897-250MG
A276897-5G
77153-742EA
15
USD
77153-742
77153-744
77153-746
77153-748
L(+)-Penicillamine 95%
L(+)-Penicillamine
Precaución: for Research Use Only
Formula:
C₅H₁₁NO₂S |
MDL Number:
MFCD00064303 CAS Number: 1113-41-3 UN: 3077 ADR: 9,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...