4-Methyl-7H-pyrrolo[2,3-d]pyrimidine 97%

Supplier: Ambeed
Warning

A276816-100MG A276816-1G A276816-250MG
77153-736EA 8.24 USD
77153-736 77153-738 77153-740
4-Methyl-7H-pyrrolo[2,3-d]pyrimidine 97%
4-Methyl-7H-pyrrolo[2,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₇H₇N₃
MDL Number: MFCD13619883
CAS Number: 945950-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR