4-(5-{4-[(Pent-1-yl)oxy]phenyl}isoxazol-3-yl)benzoic acid 95%

Supplier: Ambeed
Warning

Synonyms: 4-(5-(4-(Pentyloxy)phenyl)isoxazol-3-yl)benzoic acid, 4-{5-[4-(Pentyloxy)phenyl]-1,2-oxazol-3-yl}benzoic acid, 3-(4-Carboxyphenyl)-5-[4-(pentyloxy)phenyl]-1,2-oxazole

A276666-1G A276666-250MG A276666-5G
77153-730EA 39 USD
77153-730 77153-732 77892-541
4-(5-{4-[(Pent-1-yl)oxy]phenyl}isoxazol-3-yl)benzoic acid 95%
4-(5-{4-[(Pent-1-yl)oxy]phenyl}isoxazol-3-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₂₁H₂₁NO₄
MDL Number: MFCD20267356
CAS Number: 179162-55-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR