Methyltrioctylammonium bis(trifluoromethylsulphonyl)imide 98%

Supplier: Ambeed
Warning

Synonyms: N-Methyl-N,N-dioctyloctan-1-aminium bis((trifluoromethyl)sulphonyl)amide

A276031-100G A276031-1G A276031-25G A276031-5G
77153-722EA 353 USD
77153-722 77153-724 77153-726 77153-728
Methyltrioctylammonium bis(trifluoromethylsulphonyl)imide 98%
Methyltrioctylammonium bis(trifluoromethylsulphonyl)imide

Precaución: for Research Use Only

Formula: C₂₇H₅₄F₆N₂O₄S₂
MDL Number: MFCD08275364
CAS Number: 375395-33-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR