Toluene-4-sulfonic acid oxetan-2-yl methyl ester 98%

Supplier: Ambeed
Warning

A275906-100MG A275906-1G A275906-250MG A275906-5G
77153-714EA 41 USD
77153-714 77153-716 77153-718 77153-720
Toluene-4-sulfonic acid oxetan-2-yl methyl ester 98%
Toluene-4-sulfonic acid oxetan-2-yl methyl ester

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₄S
MDL Number: MFCD08544404
CAS Number: 115845-51-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR