N,N'-((1R,2R)-cyclohexane-1,2-diyl)dimethanesulfonamide 97%

Supplier: Ambeed
Warning

A275393-250MG A275393-100MG A275393-1G
77153-708EA 44 USD
77153-708 77901-687 77900-896
N,N'-((1R,2R)-cyclohexane-1,2-diyl)dimethanesulfonamide 97%
N,N'-((1R,2R)-cyclohexane-1,2-diyl)dimethanesulfonamide

Precaución: for Research Use Only

Formula: C₈H₁₈N₂O₄S₂
MDL Number: MFCD03411210
CAS Number: 122833-58-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR