tert-Butyl 3-methyl-4,6-dihydropyrrolo[3,4-c]pyrazole-5(1H)-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A274802-100MG
A274802-1G
A274802-250MG
A274802-5G
77153-690EA
52
USD
77153-690
77153-692
77153-694
77153-696
tert-Butyl 3-methyl-4,6-dihydropyrrolo[3,4-c]pyrazole-5(1H)-carboxylate 95%
tert-Butyl 3-methyl-4,6-dihydropyrrolo[3,4-c]pyrazole-5(1H)-carboxylate
Precaución: for Research Use Only
Formula:
C₁₁H₁₇N₃O₂ |
MDL Number:
MFCD17016069 CAS Number: 951127-35-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...