2,5-Difluoro-p-toluic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2,5-Difluoro-4-methylbenzoic acid

A274352-10G A274352-1G A274352-5G
77153-678EA 96.82 USD
77153-678 77153-680 77153-682
2,5-Difluoro-p-toluic acid 97%
2,5-Difluoro-p-toluic acid

Precaución: for Research Use Only

Formula: C₈H₆F₂O₂
MW: 172.13 g/mol
MDL Number: MFCD08063230
CAS Number: 103877-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR