8-Bromochroman 95%

Supplier: Ambeed
Warning

A274125-250MG A274125-100MG
77153-676EA 142.14 USD
77153-676 77685-776
8-Bromochroman 95%
8-Bromochroman

Precaución: for Research Use Only

Formula: C₉H₉BrO
MW: 213.07 g/mol
MDL Number: MFCD09266208
CAS Number: 3722-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR