Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]copper(I) 95%

Supplier: Ambeed
Warning

Synonyms: Chloro[1,3-bis(2,6-di-i-propylphenyl)imidazol-2-ylidene]copper(I), [1,3-Bis(2,6-diisopropylphenyl)imidazol-2-ylidene]copper chloride

A273385-1G A273385-250MG A273385-25G A273385-5G
77153-658EA 37.08 USD
77153-658 77153-660 77153-662 77153-664
Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]copper(I) 95%
Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]copper(I)

Precaución: for Research Use Only

Formula: C₂₇H₃₆ClCuN₂
MW: 487.60 g/mol
MDL Number: MFCD09264276
CAS Number: 578743-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR