1-Bromo-4-(isopropylthio)benzene 96%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-4-(propan-2-ylsulphanyl)benzene, (4-Bromophenyl)(isopropyl)sulphane

A273269-250MG A273269-1G A273269-5G
77153-656EA 22 USD
77153-656 77900-814 77900-810
1-Bromo-4-(isopropylthio)benzene 96%
1-Bromo-4-(isopropylthio)benzene

Precaución: for Research Use Only

Formula: C₉H₁₁BrS
MDL Number: MFCD20484423
CAS Number: 70398-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR