(4-Vinylphenyl)boronic acid pinacol ester 98%

Supplier: Ambeed
Warning

A269367-1G A269367-250MG A269367-25G A269367-5G
77153-608EA 42 USD
77153-608 77153-610 77153-612 77153-614
(4-Vinylphenyl)boronic acid pinacol ester 98%
(4-Vinylphenyl)boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₉BO₂
MDL Number: MFCD18733870
CAS Number: 870004-04-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR