H-Gln(isopropyl)-OH 95%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Amino-5-(isopropylamino)-5-oxopentanoic acid

A268722-100MG A268722-250MG
77153-600EA 47.38 USD
77153-600 77153-602
H-Gln(isopropyl)-OH 95%
H-Gln(isopropyl)-OH

Precaución: for Research Use Only

Formula: C₈H₁₆N₂O₃
MW: 188.23 g/mol
MDL Number: MFCD00672360
CAS Number: 4311-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR