4-Fluoro-3-hydroxybenzeneboronic acid pinacol ester 95+%

Supplier: Ambeed
Warning

Synonyms: 4-Fluoro-3-hydroxyphenylboronic acid pinacol ester, 2-FLUORO-5-(4,4,5,5-TETRAMETHYL-1,3,2-DIOXABOROLAN-2-YL)PHENOL, PHENOL, 2-FLUORO-5-(4,4,5,5-TETRAMETHYL-1,3,2-DIOXABOROLAN-2-YL)-

A268475-1G A268475-250MG A268475-5G
77153-592EA 65.92 USD
77153-592 77153-594 77153-596
4-Fluoro-3-hydroxybenzeneboronic acid pinacol ester 95+%
4-Fluoro-3-hydroxybenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₆BFO₃
MW: 238.06 g/mol
MDL Number: MFCD16994403
CAS Number: 1392234-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR