1,4-Bis((1,4,8,11-tetraazacyclotetradecan-1-yl)methyl)benzene octahydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Plerixafor octaHCl

A268315-10MG A268315-1G A268315-1MG A268315-250MG A268315-50MG A268315-5MG A268315-100MG
77153-580EA 13.39 USD
77153-580 77153-582 77153-584 77153-586 77153-588 77153-590 77685-767
1,4-Bis((1,4,8,11-tetraazacyclotetradecan-1-yl)methyl)benzene octahydrochloride 98%
1,4-Bis((1,4,8,11-tetraazacyclotetradecan-1-yl)methyl)benzene octahydrochloride

Precaución: for Research Use Only

Formula: C₂₈H₆₂Cl₈N₈
MW: 794.47 g/mol
MDL Number: MFCD04974488
CAS Number: 155148-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR