Fmoc-Lys(palmitoyl)-OH 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-palmitamidohexanoic acid
A268188-5G
A268188-1G
A268188-25G
77153-578EA
89
USD
77153-578
77685-765
77892-514
Fmoc-Lys(palmitoyl)-OH 98%
Fmoc-Lys(palmitoyl)-OH
Precaución: for Research Use Only
Formula:
C₃₇H₅₄N₂O₅ |
MDL Number:
MFCD00237026 CAS Number: 201004-46-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...