Ciprofloxacin hydrochloride monohydrate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Cyclopropyl- 6-Fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic acid hydrochloride monohydrate, 1-Cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid hydrochloride monohydrate, Nanomycopulitine
A267840-100G
A267840-100MG
A267840-10G
A267840-25G
A267840-500G
77153-568EA
54
USD
77153-568
77153-570
77153-572
77153-574
77153-576
Ciprofloxacin hydrochloride monohydrate 98%
Ciprofloxacin hydrochloride monohydrate
Precaución: for Research Use Only
Formula:
C₁₇H₁₈FN₃O₃·HCl·H₂O |
MDL Number:
MFCD00242856 CAS Number: 86393-32-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...