5-Phenylthiazole-2-carbaldehyde 95%

Supplier: Ambeed
Warning

A266989-100MG A266989-250MG
77153-564EA 110.21 USD
77153-564 77153-566
5-Phenylthiazole-2-carbaldehyde 95%
5-Phenylthiazole-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₇NOS
MW: 189.23 g/mol
MDL Number: MFCD11846732
CAS Number: 862494-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR