(E/Z)-Rigosertib sodium 99%

Supplier: Ambeed
Warning

Synonyms: Sodium (E)-2-((2-methoxy-5-(((2,4,6-trimethoxystyryl)sulphonyl)methyl)phenyl)amino)acetate, Sodium (2-methoxy-5-(((2,4,6-trimethoxystyryl)sulphonyl)methyl)phenyl)glycinate, ON-01910 (sodium salt)

A266562-10MG A266562-1MG A266562-25MG A266562-50MG A266562-5MG
77153-554EA 27.81 USD
77153-554 77153-556 77153-558 77153-560 77153-562
(E/Z)-Rigosertib sodium 99%
(E/Z)-Rigosertib sodium

Precaución: for Research Use Only

Formula: C₂₁H₂₄NNaO₈S
MW: 473.47 g/mol
MDL Number: MFCD11655911
CAS Number: 1225497-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR