3-Bromo-5-chloro-2-iodopyridine 97%

Supplier: Ambeed
Warning

A266536-100MG A266536-250MG
77153-550EA 10 USD
77153-550 77153-552
3-Bromo-5-chloro-2-iodopyridine 97%
3-Bromo-5-chloro-2-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₂BrClIN
MDL Number: MFCD11847354
CAS Number: 823221-97-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR