4-Bromo-2-(trifluoromethyl)benzoyl chloride 95%

Supplier: Ambeed
Danger

A263801-10G A263801-25G A263801-5G
77153-520EA 101 USD
77153-520 77153-522 77153-524
4-Bromo-2-(trifluoromethyl)benzoyl chloride 95%
4-Bromo-2-(trifluoromethyl)benzoyl chloride

Precaución: for Research Use Only

Formula: C₈H₃BrClF₃O
MDL Number: MFCD13185394
CAS Number: 104356-17-4
UN: 1760
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR