KRN-633 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(2-Chloro-4-((6,7-dimethoxyquinazolin-4-yl)oxy)phenyl)-3-propylurea
A263491-100MG
A263491-10MG
A263491-1MG
A263491-50MG
A263491-5MG
77153-500EA
207.03
USD
77153-500
77153-502
77153-504
77153-506
77153-508
KRN-633 98%
KRN-633
Precaución: for Research Use Only
Formula:
C₂₀H₂₁ClN₄O₄ MW: 416.86 g/mol |
MDL Number:
MFCD14155620 CAS Number: 286370-15-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...