GSK 4716 98%

Supplier: Ambeed
Danger

Synonyms: 4-Hydroxy-N'-(4-isopropylbenzylidene)benzohydrazide, GSK-4716

A262201-100MG A262201-10MG A262201-1MG A262201-25MG A262201-50MG
77153-482EA 103 USD
77153-482 77153-484 77153-486 77153-488 77153-490
GSK 4716 98%
GSK 4716

Precaución: for Research Use Only

Formula: C₁₇H₁₈N₂O₂
MDL Number: MFCD00567155
CAS Number: 101574-65-6
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR