2-Amino-5-chloro-4-iodophenol 95%

Supplier: Ambeed
Warning

A261773-100MG A261773-250MG
77153-472EA 49.44 USD
77153-472 77153-474
2-Amino-5-chloro-4-iodophenol 95%
2-Amino-5-chloro-4-iodophenol

Precaución: for Research Use Only

Formula: C₆H₅ClINO
MW: 269.47 g/mol
MDL Number: MFCD27978469
CAS Number: 1037298-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR