1-Butyl-3-methylimidazoliumtrifluoromethanesulfonate 98%

Supplier: Ambeed
Danger

Synonyms: 1-Butyl-3-methylimidazolium Trifluoromethanesulphonate

A260720-10G A260720-25G A260720-5G
77153-452EA 23.69 USD
77153-452 77153-454 77153-456
1-Butyl-3-methylimidazoliumtrifluoromethanesulfonate 98%
1-Butyl-3-methylimidazoliumtrifluoromethanesulphonate

Precaución: for Research Use Only

Formula: C₉H₁₅F₃N₂O₃S
MW: 288.29 g/mol
MDL Number: MFCD03427620
CAS Number: 174899-66-2
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR