4-Bromo-2-fluorothiophenol 95%

Supplier: Ambeed
Danger

A260469-1G A260469-25G A260469-5G
77153-446EA 45.32 USD
77153-446 77153-448 77153-450
4-Bromo-2-fluorothiophenol 95%
4-Bromo-2-fluorothiophenol

Precaución: for Research Use Only

Formula: C₆H₄BrFS
MW: 207.0634 g/mol
MDL Number: MFCD09475475
CAS Number: 174414-93-8
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR