Methyl 2-hydroxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 96%
Supplier: Ambeed
|
Warning
|
A260466-1G
A260466-250MG
A260466-5G
77153-442EA
69
USD
77153-442
77153-444
77901-857
Methyl 2-hydroxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 96%
Methyl 2-hydroxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Precaución: for Research Use Only
Formula:
C₁₄H₁₉BO₅ |
MDL Number:
MFCD18730333 CAS Number: 1352730-33-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...