(S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-(3-((2,2,4,6,7-pentamethyl-2,3-dihydrobenzofuran-5-yl)sulfonyl)guanidino)hexanoic acid 96%

Supplier: Ambeed
Warning

Synonyms: Fmoc-β-HoArg(Pbf)-OH

A260439-100MG A260439-1G A260439-250MG
77153-436EA 22.66 USD
77153-436 77153-438 77153-440
(S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-(3-((2,2,4,6,7-pentamethyl-2,3-dihydrobenzofuran-5-yl)sulfonyl)guanidino)hexanoic acid 96%
(S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-(3-((2,2,4,6,7-pentamethyl-2,3-dihydrobenzofuran-5-yl)sulfonyl)guanidino)hexanoic acid

Precaución: for Research Use Only

Formula: C₃₅H₄₂N₄O₇S
MW: 662.8 g/mol
MDL Number: MFCD03490520
CAS Number: 401915-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR