Methyl-2-(bromomethyl)-5-nitrobenzoate 96%

Supplier: Ambeed
Danger

A260328-1G A260328-250MG A260328-5G A260328-25G
77153-430EA 95 USD
77153-430 77153-432 77153-434 77571-606
Methyl-2-(bromomethyl)-5-nitrobenzoate 96%
Methyl 2-(bromomethyl)-5-nitrobenzoate

Precaución: for Research Use Only

Formula: C₉H₈BrNO₄
MDL Number: MFCD04114313
CAS Number: 90725-68-1
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR