(9-([1,1'-Biphenyl]-4-yl)-9H-carbazol-3-yl)boronic acid 98%

Supplier: Ambeed
Warning

A260275-1G A260275-25G A260275-5G
77153-428EA 18 USD
77153-428 77900-724 77900-741
(9-([1,1'-Biphenyl]-4-yl)-9H-carbazol-3-yl)boronic acid 98%
(9-([1,1'-Biphenyl]-4-yl)-9H-carbazol-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₂₄H₁₈BNO₂
MDL Number: MFCD22581304
CAS Number: 1028648-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR