4-Bromo-2,3-Dichlorophenol 95%

Supplier: Ambeed
Warning

A260264-100MG A260264-1G A260264-250MG A260264-5G
77153-420EA 35.02 USD
77153-420 77153-422 77153-424 77153-426
4-Bromo-2,3-Dichlorophenol 95%
4-Bromo-2,3-Dichlorophenol

Precaución: for Research Use Only

Formula: C₆H₃BrCl₂O
MW: 241.90 g/mol
MDL Number: MFCD00152292
CAS Number: 1940-44-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR