2,6-Difluoro-3,5-dimethoxyaniline 97%

Supplier: Ambeed
Warning

A259588-100MG A259588-1G A259588-250MG A259588-25G A259588-5G
77153-398EA 7.21 USD
77153-398 77153-400 77153-402 77153-404 77153-406
2,6-Difluoro-3,5-dimethoxyaniline 97%
2,6-Difluoro-3,5-dimethoxyaniline

Precaución: for Research Use Only

Formula: C₈H₉F₂NO₂
MW: 189.159 g/mol
MDL Number: MFCD12973804
CAS Number: 651734-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR