5-bromoquinolin-6-amine 95%

Supplier: Ambeed
Warning

A258720-100MG A258720-1G A258720-250MG A258720-25G A258720-5G
77153-376EA 53 USD
77153-376 77153-378 77153-380 77153-382 77153-384
5-bromoquinolin-6-amine 95%
5-bromoquinolin-6-amine

Precaución: for Research Use Only

Formula: C₉H₇BrN₂
MDL Number: MFCD00234503
CAS Number: 50358-42-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR