AZD-8931 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4-((4-((3-Chloro-2-fluorophenyl)amino)-7-methoxyquinazolin-6-yl)oxy)piperidin-1-yl)-N-methylacetamide, Momelotinib, 4-(3-Chloro-2-fluoroanilino)-7-methoxy-6-[[1-(N-methylcarbamoylmethyl)piperidin-4-yl]oxy]quinazoline, Sapitinib

A258461-100MG A258461-10MG A258461-1MG A258461-25MG A258461-50MG A258461-5MG
77153-364EA 125.66 USD
77153-364 77153-366 77153-368 77153-370 77153-372 77153-374
AZD-8931 98%
AZD-8931

Precaución: for Research Use Only

Formula: C₂₃H₂₅ClFN₅O₃
MW: 473.93 g/mol
MDL Number: MFCD18385011
CAS Number: 848942-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR