2-Iodo-5-(trifluoromethyl)benzonitrile 98%
Supplier: Ambeed
|
Warning
|
A258225-1G
A258225-5G
77153-360EA
11
USD
77153-360
77153-362
2-Iodo-5-(trifluoromethyl)benzonitrile 98%
2-Iodo-5-(trifluoromethyl)benzonitrile
Precaución: for Research Use Only
Formula:
C₈H₃F₃IN |
MDL Number:
MFCD18398159 CAS Number: 868166-20-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...