2,5-Difluoropyridine-4-boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (2,5-Difluoropyridin-4-yl)boronic acid

A258050-1G A258050-250MG A258050-5G
77153-354EA 153.47 USD
77153-354 77153-356 77153-358
2,5-Difluoropyridine-4-boronic acid 97%
2,5-Difluoropyridine-4-boronic acid

Precaución: for Research Use Only

Formula: C₅H₄BF₂NO₂
MW: 158.899 g/mol
MDL Number: MFCD16556243
CAS Number: 1263375-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR