(1S,2S,5R)-rel-3-Azabicyclo[3.1.0]hexan-2-ylmethanol hydrochloride 98+%

Supplier: Ambeed
Warning

A256553-1G A256553-50MG
77153-348EA 1265.87 USD
77153-348 77153-350
(1S,2S,5R)-rel-3-Azabicyclo[3.1.0]hexan-2-ylmethanol hydrochloride 98+%
(1S,2S,5R)-rel-3-Azabicyclo[3.1.0]hexan-2-ylmethanol hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₂ClNO
MW: 149.61 g/mol
MDL Number: MFCD29037490
CAS Number: 1818847-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR