Trimethoxy(3-methoxypropyl)silane 95%

Supplier: Ambeed
Danger

A255792-25G A255792-5G
77153-332EA 38.11 USD
77153-332 77153-334
Trimethoxy(3-methoxypropyl)silane 95%
Trimethoxy(3-methoxypropyl)silane

Precaución: for Research Use Only

Formula: C₇H₁₈O₄Si
MW: 194.30 g/mol
MDL Number: MFCD00054721
CAS Number: 33580-59-5
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR