Pyrene-4,5,9,10-tetraone 98%

Supplier: Ambeed
Warning

A255507-100MG A255507-1G A255507-250MG A255507-5G
77153-318EA 50.47 USD
77153-318 77153-320 77153-322 77568-480
Pyrene-4,5,9,10-tetraone 98%
Pyrene-4,5,9,10-tetraone

Precaución: For research use only.

Formula: C₁₆H₆O₄
MW: 262.21 g/mol
MDL Number: MFCD11502676
CAS Number: 14727-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR