BAY 63-2521 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl (4,6-diamino-2-(1-(2-fluorobenzyl)-1H-pyrazolo[3,4-b]pyridin-3-yl)pyrimidin-5-yl)(methyl)carbamate, Riociguat
A255332-10MG
A255332-1MG
A255332-50MG
A255332-5MG
A255332-1G
A255332-250MG
A255332-100MG
77153-310EA
14.42
USD
77153-310
77153-312
77153-314
77153-316
77612-450
77612-452
77685-725
BAY 63-2521 98%
BAY 63-2521
Precaución: for Research Use Only
Formula:
C₂₀H₁₉FN₈O₂ MW: 422.42 g/mol |
MDL Number:
MFCD19443708 CAS Number: 625115-55-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...