Ethyl-2,4,5-trifluorobenzoylacetate 97%

Supplier: Ambeed
Warning

A254526-100G A254526-10G A254526-25G A254526-5G
77153-300EA 203 USD
77153-300 77153-302 77153-304 77153-306 77153-308
Ethyl-2,4,5-trifluorobenzoylacetate 97%
Ethyl-2,4,5-trifluorobenzoylacetate

Precaución: for Research Use Only

Formula: C₁₁H₉F₃O₃
MDL Number: MFCD00792431
CAS Number: 98349-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR