Methyl 8-bromo-2,3-dihydrobenzo[b][1,4]dioxine-5-carboxylate 95%

Supplier: Ambeed
Warning

A254206-100MG A254206-250MG A254206-25MG
77153-286EA 300.76 USD
77153-286 77153-288 77153-290
Methyl 8-bromo-2,3-dihydrobenzo[b][1,4]dioxine-5-carboxylate 95%
Methyl 8-bromo-2,3-dihydrobenzo[b][1,4]dioxine-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉BrO₄
MW: 273.08 g/mol
MDL Number: MFCD29919264
CAS Number: 823225-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR