1-[4-(Diphenylmethyl)-1-piperazinyl]-3,3-diphenyl-1-propanone 98%

Supplier: Ambeed
Warning

A254092-100MG A254092-1MG A254092-250MG A254092-50MG
77153-278EA 75.19 USD
77153-278 77153-280 77153-282 77153-284
1-[4-(Diphenylmethyl)-1-piperazinyl]-3,3-diphenyl-1-propanone 98%
1-[4-(Diphenylmethyl)-1-piperazinyl]-3,3-diphenyl-1-propanone

Precaución: for Research Use Only

Formula: C₃₂H₃₂N₂O
MW: 460.60 g/mol
MDL Number: MFCD03387795
CAS Number: 41332-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR